missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dextran Sulfate Sodium Salt, Thermo Scientific Chemicals
£147.00 - £666.00
Chemical Identifiers
CAS | 9011-18-1 |
---|---|
Molecular Formula | (C6H7O5)m(C6H7O5)n |
Molecular Weight (g/mol) | 40,000 |
MDL Number | MFCD00081551 |
InChI Key | BLFLLBZGZJTVJG-UHFFFAOYSA-N |
Synonym | benzocaine, ethyl aminobenzoate, ethyl p-aminobenzoate, americaine, anesthesin, anaesthesin, ethoform, norcaine, orthesin, parathesin |
PubChem CID | 2337 |
ChEBI | CHEBI:116735 |
IUPAC Name | ethyl 4-aminobenzoate |
SMILES | *-OC[C@H]1O[C@H](OC[C@H]2O[C@H](-*)[C@H](O[*])[C@@H](O[*])[C@@H]2O[*])[C@H](O[*])[C@@H](O-*)[C@@H]1O[*] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15875118
|
Thermo Scientific
J14489.09 |
10 g | Plastic bottle |
£147.00
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15885118
|
Thermo Scientific
J14489.18 |
50 g | Plastic bottle |
£441.00
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15895118
|
Thermo Scientific
J14489.22 |
100 g | Plastic bottle |
£666.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
9011-18-1 | |
40,000 | |
BLFLLBZGZJTVJG-UHFFFAOYSA-N | |
2337 | |
ethyl 4-aminobenzoate |
(C6H7O5)m(C6H7O5)n | |
MFCD00081551 | |
benzocaine, ethyl aminobenzoate, ethyl p-aminobenzoate, americaine, anesthesin, anaesthesin, ethoform, norcaine, orthesin, parathesin | |
CHEBI:116735 | |
*-OC[C@H]1O[C@H](OC[C@H]2O[C@H](-*)[C@H](O[*])[C@@H](O[*])[C@@H]2O[*])[C@H](O[*])[C@@H](O-*)[C@@H]1O[*] |
Specifications
9011-18-1 | |
(C6H7O5)m(C6H7O5)n | |
benzocaine, ethyl aminobenzoate, ethyl p-aminobenzoate, americaine, anesthesin, anaesthesin, ethoform, norcaine, orthesin, parathesin | |
*-OC[C@H]1O[C@H](OC[C@H]2O[C@H](-*)[C@H](O[*])[C@@H](O[*])[C@@H]2O[*])[C@H](O[*])[C@@H](O-*)[C@@H]1O[*] | |
40,000 | |
CHEBI:116735 | |
Dextran Sulfate Sodium Salt |
Powder | |
MFCD00081551 | |
BLFLLBZGZJTVJG-UHFFFAOYSA-N | |
ethyl 4-aminobenzoate | |
2337 | |
40,000- 50,000 |
Safety and Handling
missing translation for 'rtecsNumber' : HH9290000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Dextran Sulfate Sodium Salt, Thermo Scientific Chemicals