missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Dextrin Sepharose™ High Performance Media

Provide convenient affinity purification of recombinant proteins with maltose binding protein (MBP) tags

Brand:  Cytiva 28-9355-98

Code : GE

Additional Details : Weight : 0.31200kg


 View more versions of this product

Product Code. 10566305

  • £2524.00 / 100mL
Estimated Shipment: 09-09-2024
to see stock.
Add to basket

Description

Description

  • Physiological conditions and mild elution preserves activity of target protein
  • Robust, stable medium allows narrow peaks in elution of target protein
  • Compatible with commonly used aqueous buffers and easily regenerated with 0.5M NaOH
  • Easy scale-up

Specifications

Specifications

100 mL
Dextrin Sepharose
34 μm
Affinity Resin
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Dextrin Sepharose High Performance, 100 ml

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P235-Keep cool.
  • P501-Dispose of contents/container to .

Supplemental information
  • MIXTURE LIST-Contains : ethanol

SDS
Documents

Documents

Certificates
Special Offers

Special Offers