missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dextrin, Thermo Scientific Chemicals
£22.81 - £57.89
Chemical Identifiers
CAS | 9004-53-9 |
---|---|
Molecular Formula | C18H32O16 |
Molecular Weight (g/mol) | 504.438 |
MDL Number | MFCD00081554 |
InChI Key | FYGDTMLNYKFZSV-MRCIVHHJSA-N |
Synonym | dextrin, dextrins, fortodex, dextrid, corn dextrin, dextrina bianca, british gum, crystal gum, dextrin 3, caloreen |
PubChem CID | 62698 |
IUPAC Name | (3R,4S,5S,6R)-2-[(2R,3S,4R,5R)-4,5-dihydroxy-2-(hydroxymethyl)-6-[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxan-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol |
SMILES | C(C1C(C(C(C(O1)OC2C(OC(C(C2O)O)OC3C(OC(C(C3O)O)O)CO)CO)O)O)O)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10187030
|
Thermo Scientific Acros
406281000 |
100 g | Plastic bottle |
£22.81
100g |
Estimated Shipment: 18-11-2024
Log in to see stock. |
||||
10351441
|
Thermo Scientific Acros
406285000 |
500 g | Plastic bottle |
£57.89
500g |
In Stock
Log in to see stock. |
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
9004-53-9 | |
504.438 | |
FYGDTMLNYKFZSV-MRCIVHHJSA-N | |
62698 | |
C(C1C(C(C(C(O1)OC2C(OC(C(C2O)O)OC3C(OC(C(C3O)O)O)CO)CO)O)O)O)O |
C18H32O16 | |
MFCD00081554 | |
dextrin, dextrins, fortodex, dextrid, corn dextrin, dextrina bianca, british gum, crystal gum, dextrin 3, caloreen | |
(3R,4S,5S,6R)-2-[(2R,3S,4R,5R)-4,5-dihydroxy-2-(hydroxymethyl)-6-[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxan-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol |
Specifications
White to Yellow | |
Crystalline Powder | |
(C6H10O5)x | |
15, 2955 | |
Solubility in water: soluble in hot water. Other solubilities: insoluble in ether and alcohols | |
C(C1C(C(C(C(O1)OC2C(OC(C(C2O)O)OC3C(OC(C(C3O)O)O)CO)CO)O)O)O)O | |
504.438 | |
162.07 | |
Dextrin |
9004-53-9 | |
C18H32O16 | |
MFCD00081554 | |
dextrin, dextrins, fortodex, dextrid, corn dextrin, dextrina bianca, british gum, crystal gum, dextrin 3, caloreen | |
FYGDTMLNYKFZSV-MRCIVHHJSA-N | |
(3R,4S,5S,6R)-2-[(2R,3S,4R,5R)-4,5-dihydroxy-2-(hydroxymethyl)-6-[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxan-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol | |
62698 | |
Authentic |
Safety and Handling
missing translation for 'einecsNumber' : 232-675-4
missing translation for 'rtecsNumber' : HH9450000
missing translation for 'tsca' : TSCA
RUO – Research Use Only