missing translation for 'onlineSavingsMsg'
Learn More
Di-p-toluoyl-D-tartaric acid, 98%, Thermo Scientific Chemicals Product Code.: 11327976

Di-p-toluoyl-D-tartaric acid, 98%, Thermo Scientific Chemicals

Product Code. 11327976
25 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11327976

Brand: Thermo Scientific Alfa Aesar L13223.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32634-68-7
Molecular Formula C20H18O8
Molecular Weight (g/mol) 386.36
MDL Number MFCD00008552
InChI Key CMIBUZBMZCBCAT-HOTGVXAUSA-N
Synonym di-p-toluoyl-d-tartaric acid, +-di-1,4-toluoyl-d-tartaric acid, 2s,3s-2,3-bis 4-methylbenzoyl oxy succinic acid, unii-4480ma5qip, ditoluoyltartaric acid, +, +-dtta, butanedioic acid, 2,3-bis 4-methylbenzoyl oxy-, 2s,3s, 2s,3s-2,3-bis 4-methylbenzoyl oxy butanedioic acid, --di-p-toluoyl-l-trataric acidShow MoreShow Less
PubChem CID 263211
SMILES CC1=CC=C(C=C1)C(=O)O[C@@H]([C@H](OC(=O)C1=CC=C(C)C=C1)C(O)=O)C(O)=O

Specifications

Melting Point 169°C to 171°C
Beilstein 3225584
IUPAC Name (2S,3S)-2,3-bis(4-methylbenzoyloxy)butanedioic acid
Formula Weight 386.36
Percent Purity 98%
Quantity 25 g
Optical Rotation +139° (c=1 in Ethanol)
Chemical Name or Material Di-p-toluoyl-D-tartaric acid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberD30Y001 Certificate TypeCertificate of Analysis Date19/03/2025 Catalog NumberL13223.22,L13223.06,L13223.14
Lot NumberU23K017 Certificate TypeCertificate of Analysis Date11/09/2024 Catalog NumberL13223.22,L13223.14
Lot Number10172289 Certificate TypeCertificate of Analysis Date05/09/2023 Catalog NumberL13223.22,L13223.14
Lot NumberP16J001 Certificate TypeCertificate of Analysis Date09/05/2023 Catalog NumberL13223.22,L13223.14
Lot NumberN17J030 Certificate TypeCertificate of Analysis Date09/05/2023 Catalog NumberL13223.22,L13223.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Di-p-toluoyl-D-tartaric acid, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.