missing translation for 'onlineSavingsMsg'
Learn More
Di-Sodium Hydrogen Orthophosphate Dodecahydrate, Certified AR for Analysis, meets analytical specification of Ph.Eur., BP, Fisher Chemical™ Product Code.: 10204020

Di-Sodium Hydrogen Orthophosphate Dodecahydrate, Certified AR for Analysis, meets analytical specification of Ph.Eur., BP, Fisher Chemical™

Product Code. 10204020
1 kg
Click to view available options
Quantity:
500 g
1 kg
This item is not returnable. View return policy

Product Code. 10204020

Brand: Fisher Chemical S/4400/60

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Di-Sodium Hydrogen Orthophosphate Dodecahydrate, Certified AR for Analysis, meets analytical specification of Ph.Eur., BP, Fisher Chemical, Quantity: 500g, CAS: 10039-32-4, Melting Point: 35 deg.C, Molecular Formula: H25Na2O16P, Molecular Weight: 358.137, Packaging: Plastic powder jar

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10039-32-4
Molecular Formula H25Na2O16P
Molecular Weight (g/mol) 358.137
MDL Number 149181
InChI Key DGLRDKLJZLEJCY-UHFFFAOYSA-L
Synonym disodium phosphate dodecahydrate, disodium hydrogen phosphate dodecahydrate, phosphoric acid, disodium salt, dodecahydrate, di-sodium hydrogen phosphate dodecahydrate, sodium hydrogenphosphate dodecahydrate, unii-e1w4n241fo, sodium phosphate dibasic dodecahydrate, sodium phosphate, dibasic, dodecahydrate, disodium orthophosphate, dodecahydrate, sodium monohydrogen phosphate dodecahydrate 2:1:1:12Show MoreShow Less
PubChem CID 61456
IUPAC Name disodium;hydrogen phosphate;dodecahydrate
SMILES O.O.O.O.O.O.O.O.O.O.O.O.OP(=O)([O-])[O-].[Na+].[Na+]

Specifications

Melting Point 35°C
pH 9.0 to 9.4
Physical Form Solid
Quantity 1 kg
Formula Weight 358.13g/mol
Packaging Plastic powder jar
  • Multiple Manufacturers
    Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.
    Promo Code: 24155

    Expires2025-04-30

    Days

    29

    Hours

    15

    Minutes

    17

    Seconds

    36
    Hot Deal

    *Discount will apply to qualifying orders received by Fisher Scientific between 01/03/2025 and 31/03/2025. Promotion code must be presented at the time of order. Discount applies to the list price in effect at the time the order is received. Promotion cannot be combined with other discounts or promotions. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy. Other restrictions may apply.

    Terms & Conditions See Full Promo Details
  • Multiple Manufacturers
    Shop now and save up to 97% on top-quality products for your lab needs.
    Promo Code: 25908

    Expires2025-03-31

    Days

    0

    Hours

    8

    Minutes

    42

    Seconds

    23
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Di-Sodium Hydrogen Orthophosphate Dodecahydrate, Certified AR for Analysis, meets analytical specification of Ph.Eur., BP, Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.