missing translation for 'onlineSavingsMsg'
Learn More
Di-tert-butyl hydrazodiformate, 97%, Thermo Scientific Chemicals Product Code.: 11955671

Di-tert-butyl hydrazodiformate, 97%, Thermo Scientific Chemicals

Product Code. 11955671
10 g, Glass bottle
Click to view available options
Quantity:
10 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 11955671

Brand: Thermo Scientific Acros 440420100

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 16466-61-8
Melting Point 120.0°C to 125.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Molecular Formula C10H20N2O4
Quantity 10 g
Synonym di-tert-butyl hydrazodicarboxylate, di-tert-butyl hydrazine-1,2-dicarboxylate, di-tert-butyl hydrazodiformate, di-tert-butyl bicarbamate, n,n'-di-boc-hydrazine, n,n'-bis tert-butoxycarbonyl hydrazine, di-tert-butyl-1,2-hydrazodicarboxylate, 1,2-hydrazinedicarboxylic acid, bis 1,1-dimethylethyl ester, tert-butyl n-2-methylpropan-2-yl oxycarbonylamino carbamate, n'-tert-butoxy carbonyl tert-butoxy carbohydrazide
InChI Key TYSZETYVESRFNT-UHFFFAOYSA-N
SMILES CC(C)(C)OC(=O)NNC(=O)OC(C)(C)C
IUPAC Name tert-butyl N-[(2-methylpropan-2-yl)oxycarbonylamino]carbamate
Molecular Weight (g/mol) 232.28
PubChem CID 85431
Formula Weight 232.28
Percent Purity 97%
Physical Form Crystalline Powder or Powder
Chemical Name or Material Di-tert-butyl hydrazodiformate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Catalog Number
Lot Number343686 Certificate TypeCertificate of Analysis Date08/02/2023 Catalog Number440420100
Lot Number343686 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number440420100
compliance-icons
Product Identifier
  • Di-tert-butyl hydrazodiformate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Di-tert-butyl hydrazodiformate, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.