Learn More
MP Biomedicals Diagnostic Kit for Luteinizing Hormone (LH) (MPQuanti™ only)
Diagnostic Kit for Luteinizing Hormone is intended for in-vitro quantitative determination of Luteinizing Hormone (LH) in human whole blood, serum, and plasma.
Brand: MP Biomedicals 07LH5025
This item is not returnable.
View return policy
Description
- Diagnostic Kit for LH (Whole blood/Serum/Plasma) is intended for in vitro quantitative determination of luteinizing hormone (LH) in human whole blood/serum/plasma as an aid in the detection of ovulation.
- The specimen moves through the strip from specimen pad to absorbent pad. If the specimen contains LH, it attaches to the fluorescent microspheres-conjugated anti-LH antibodies.
- Then the complex will be captured by the capture antibodies coated on the nitrocellulose membrane (Test line).
- The concentration of LH in the specimen correlates linearly with the fluorescence signal intensity captured on the T line.
- According to the fluorescence intensity of the test and standard curve, the concentration of LH in the specimen is calculated by the MPQuanti™ Immuflorescence Analyzer.
- CE marked
- For professional in-vitro diagnostic use
- Working only with MPQuanti™ Immunofluorescence Analyzer
- Linear working range: 1-300 mIU/mL, R≥0.990
- Test results should be interpreted at a few seconds (QUICK TEST) or 15 minutes (STANDART TEST)
Specifications
LH Surge (Luteinizing Hormone or hLH) | |
Human whole blood, Human serum, Human plasma | |
Ready-to-Use (RTU) |
Immunofluorescence | |
POC, Quantitative, LH, hormone |
Product Suggestions
Customers who viewed this item also viewed.
Product Content Correction
Product Title
MP Biomedicals Diagnostic Kit for Luteinizing Hormone (LH) (MPQuanti™ only) > LH
Spot an opportunity for improvement?Share a Content Correction