Learn More
MP Biomedicals Diagnostic Kit for Triiodothyronine (T3) (MPQuanti™ only)
Diagnostic Kit for Triiodothyronine (T3) is intended for in-vitro quantitative detection of total Triiodothyronine (T3) in human serum or plasma.
Brand: MP Biomedicals 07T35025
This item is not returnable.
View return policy
Description
- Diagnostic Kit for Triiodothyronine (T3) (Serum/Plasma) is based on fluorescence immunoassay technology.
- The specimen moves through the strip from sample pad to absorbent pad. T3 in the specimen will compete with the T3 antigen coated on the strip.
- The less T3 in the specimen, the more fluorescent microspheres conjugated with anti-T3 antibodies can be captured by the T3 antigen coated on the strip.
- The concentration of T3 in the sample is inversely related to the intensity of the fluorescent signal captured on the T line.
- According to the fluorescence intensity of the test and the standard curve, the concentration of T3 in the sample is calculated by the, compatible only with the MPQuanti™ Immuflorescence Analyzer, to show T3 concentration in the sample.
- CE marked
- For professional in-vitro diagnostic use
- Working only with MPQuanti™ Immunofluorescence Analyzer
- Linear working range: 62 to 9.24nmol/L, R≥0.990
- Test results should be interpreted at 15 minutes
Specifications
Total T3 | |
Human serum, Human plasma | |
Ready-to-Use (RTU) |
Immunofluorescence | |
POC, Quantitative, T3, Triiodothyronine |
Product Suggestions
Customers who viewed this item also viewed
Product Content Correction
Product Title
MP Biomedicals Diagnostic Kit for Triiodothyronine (T3) (MPQuanti™ only) > T3
Spot an opportunity for improvement?Share a Content Correction