missing translation for 'onlineSavingsMsg'
Learn More
Diatomaceous Earth, J.T. Baker™ Product Code.: 15538774

Diatomaceous Earth, J.T. Baker™

Product Code. 15538774
1 kg, Poly Pail
Click to view available options
Quantity:
500 g
1 kg
Packaging:
Poly Pail
Wide Mouth Amber Glass Bottle
This item is not returnable. View return policy

Product Code. 15538774

Brand: Avantor J.T.Baker E40608

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 68855-54-9
Molecular Formula CNa2O3
Molecular Weight (g/mol) 105.988
InChI Key CDBYLPFSWZWCQE-UHFFFAOYSA-L
PubChem CID 10340
ChEBI CHEBI:29377
IUPAC Name disodium;carbonate
SMILES C(=O)([O-])[O-].[Na+].[Na+]

Specifications

Packaging Poly Pail
Particle Size 33 Micron
Specific Gravity 1L = 2.30kg
Quantity 1 kg
Chemical Name or Material Diatomaceous Earth
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Diatomaceous Earth, J.T. Baker™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.