Learn More
Dibutylboron trifluoromethanesulfonate, 1M sol. in dichloromethane, Thermo Scientific Chemicals
Brand: Thermo Scientific Acros 378991000
111.65 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.
View more versions of this product
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acidSpecifications
Dibutylboron trifluoromethanesulfonate | |
Orange to Yellow | |
27°C | |
Glass Bottle | |
CF3SO3B[(CH2)3CH3]2 | |
100 mL | |
1.271 | |
Solubility in water: reacts. | |
CCCCB(CCCC)OS(=O)(=O)C(F)(F)F | |
274.11 | |
274.11 |
60669-69-4, 75-09-2 | |
1.2710g/mL | |
98% | |
C9H18BF3O3S | |
MFCD00009669 | |
07,91; 09,140; 16,109 | |
dibutylboron trifluoromethanesulfonate, dibutylboryl trifluoromethanesulfonate solution, nbu2botf, bu2botf, dibutylboron triflate, n-bu 2botf, dibutylboryl trifluoromethanesulfonate, dibutylboron triflate solution, trifluoromethylsulfonyloxy dibutylborane, dibutyl trifluoromethyl sulfonyl oxy borane | |
FAVAVMFXAKZTMV-UHFFFAOYSA-N | |
dibutylboranyl trifluoromethanesulfonate | |
2724243 | |
Solution |
Safety and Handling
- Dibutylboron trifluoromethanesulfonate in dichloromethane
Signal Word
- Danger
Hazard Category
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 3
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity Category 3
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H351-Suspected of causing cancer.
- H336-May cause drowsiness or dizziness.
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only