Learn More
Dibutylboron trifluoromethanesulfonate, 1M sol. in dichloromethane, Thermo Scientific Chemicals
£52.26 - £147.74
Specifications
Chemical Name or Material | Dibutylboron trifluoromethanesulfonate |
---|---|
CAS | 60669-69-4, 75-09-2 |
Color | Orange to Yellow |
Density | 1.2710g/mL |
Flash Point | 27°C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10297013
|
Thermo Scientific Acros
378990250 |
25 mL | Glass Bottle |
£52.26
25mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Currently Unavailable: We have no information when this product will be available
View alternative products
|
10210493
|
Thermo Scientific Acros
378991000 |
100 mL | Glass Bottle |
£147.74
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acidSpecifications
Dibutylboron trifluoromethanesulfonate | |
Orange to Yellow | |
27°C | |
Glass Bottle | |
CF3SO3B[(CH2)3CH3]2 | |
07,91; 09,140; 16,109 | |
dibutylboron trifluoromethanesulfonate, dibutylboryl trifluoromethanesulfonate solution, nbu2botf, bu2botf, dibutylboron triflate, n-bu 2botf, dibutylboryl trifluoromethanesulfonate, dibutylboron triflate solution, trifluoromethylsulfonyloxy dibutylborane, dibutyl trifluoromethyl sulfonyl oxy borane | |
FAVAVMFXAKZTMV-UHFFFAOYSA-N | |
dibutylboranyl trifluoromethanesulfonate | |
2724243 | |
Solution |
60669-69-4, 75-09-2 | |
1.2710g/mL | |
98% | |
C9H18BF3O3S | |
MFCD00009669 | |
1.271 | |
Solubility in water: reacts. | |
CCCCB(CCCC)OS(=O)(=O)C(F)(F)F | |
274.11 | |
274.11 |
Safety and Handling
GHS H Statement
Flammable liquid and vapour.
Suspected of causing cancer.
Causes severe skin burns and eye damage.
May cause drowsiness or dizziness.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse
GHS Signal Word: Danger
- Dibutylboron trifluoromethanesulfonate in dichloromethane
Signal Word
- Danger
Hazard Category
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 3
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity Category 3
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H351-Suspected of causing cancer.
- H336-May cause drowsiness or dizziness.
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only