missing translation for 'onlineSavingsMsg'
Learn More

Dichloromethane, 99.8+%, for analysis, stabilized with amylene, Thermo Scientific Chemicals

£20.07 - £542.70

Chemical Identifiers

CAS 75-09-2, 513-35-9
Molecular Formula CH2Cl2
Molecular Weight (g/mol) 84.93
MDL Number MFCD00000881
InChI Key YMWUJEATGCHHMB-UHFFFAOYSA-N
Synonym methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mmShow MoreShow Less
PubChem CID 6344
ChEBI CHEBI:15767
IUPAC Name dichloromethane
SMILES ClCCl
View More Specs

Products 5
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10785941
SDS View Documents Promotion Details
Thermo Scientific Acros
113460010
1 L Glass Bottle
£20.07
1 L
Please to purchase this item. Need a web account? Register with us today!
10396410
SDS View Documents Promotion Details
Thermo Scientific Acros
113460025
2.5 L Glass Bottle
£29.88
2.50 L
Please to purchase this item. Need a web account? Register with us today!
10149082
SDS View Documents
Thermo Scientific Acros
113460050
5 L Plastic Drum This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10343013
SDS View Documents
Thermo Scientific Acros
113460100
10 L Metal Drum
£223.11
10 L
Please to purchase this item. Need a web account? Register with us today!
10162330
SDS View Documents
Thermo Scientific Acros
113460250
25 L Metal Drum
£542.70
25 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For analysis
Specifications
Show More
Videos
Safety and Handling
Show More
SDS
Documents

Documents

Certificates

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Dichloromethane, 99.8+%, for analysis, stabilized with amylene, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.