missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dichloromethane, Biotech Grade, 99.9%, Honeywell Burdick & Jackson™
Biotech Grade, 99.9%, contains 40-150 ppm amylene as stabilizer
£62.67 - £112.42
Chemical Identifiers
CAS | 75-09-2 |
---|---|
Molecular Formula | CH2Cl2 |
Molecular Weight (g/mol) | 84.93 |
MDL Number | MFCD00000881 |
InChI Key | YMWUJEATGCHHMB-UHFFFAOYSA-N |
Synonym | methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm |
PubChem CID | 6344 |
ChEBI | CHEBI:15767 |
IUPAC Name | dichloromethane |
SMILES | ClCCl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15655670
|
Honeywell-Burdick and Jackson
494453-1L |
1 L | Glass Bottle |
£62.67
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15665670
|
Honeywell-Burdick and Jackson
494453-2L |
2 L | Glass Bottle |
£112.42
2 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Honeywell Burdick & Jackson™ (B&J) developed the first high-purity solvents more than 50 years ago
- As new applications and techniques emerge, Burdick & Jackson continues to develop products to meet the most demanding requirements for laboratories and pharmaceutical production
- Our packaging solutions, ranging from lab scale through production, provide our customers with added value, increased efficiency, and enhanced safety.
Chemical Identifiers
75-09-2 | |
84.93 | |
YMWUJEATGCHHMB-UHFFFAOYSA-N | |
6344 | |
dichloromethane |
CH2Cl2 | |
MFCD00000881 | |
methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm | |
CHEBI:15767 | |
ClCCl |
Specifications
-97°C | |
Colorless | |
39°C to 41°C | |
1223°F | |
Liquid | |
MFCD00000881 | |
UN1593 | |
methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm | |
ClCCl | |
84.93 | |
CHEBI:15767 | |
99.90% |
75-09-2 | |
1.325g/mL at 25°C | |
0.22 | |
Sweet | |
CH2Cl2 | |
24.45psi (55°C), 6.83psi (20°C) | |
1730800 | |
YMWUJEATGCHHMB-UHFFFAOYSA-N | |
dichloromethane | |
6344 | |
84.93g/mol | |
Dichloromethane |
Safety and Handling
P261-P281-P305 + P351 + P338
missing translation for 'dotInformation' : 6.1/PGIII
missing translation for 'rtecsNumber' : 200-838-9