missing translation for 'onlineSavingsMsg'
Learn More

Dichloromethane-d{2}, 99.9% (Isotopic), Thermo Scientific Chemicals

£30.95 - £139.70

Chemical Identifiers

CAS 1665-00-5
Molecular Formula CH2Cl2
Molecular Weight (g/mol) 86.939
MDL Number MFCD00000882
InChI Key YMWUJEATGCHHMB-DICFDUPASA-N
Synonym dichloromethane-d2, methylene chloride-d2, dichloro 2h2 methane, methane-d2, dichloro, cd2cl2, deuterated dichloromethane, dichloro dideuterio methane, dideuteromethylene chloride, dichloromethane-d2, 99.9 atom % d, dichloromethane-d', 99.96 atom % d
PubChem CID 160586
IUPAC Name dichloro(dideuterio)methane
SMILES C(Cl)Cl
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11357958
View Documents
Thermo Scientific Alfa Aesar
036500.03
1 g
£30.95
1g
Please to purchase this item. Need a web account? Register with us today!
11367958
View Documents
Thermo Scientific Alfa Aesar
036500.06
5 g
£139.70
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

In Anion-templated assembly of [2] rotaxanes, the addition of tetrabutylammonium (TBA) halide, dihydrogen phosphate and hydrogen sulfate salts to dichloromethane-d 2 solutions of both derivatives resulted in significant downfield shift perturbations of the respective compounds' aryl b and amide c protons.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1665-00-5
86.939
YMWUJEATGCHHMB-DICFDUPASA-N
160586
C(Cl)Cl
CH2Cl2
MFCD00000882
dichloromethane-d2, methylene chloride-d2, dichloro 2h2 methane, methane-d2, dichloro, cd2cl2, deuterated dichloromethane, dichloro dideuterio methane, dideuteromethylene chloride, dichloromethane-d2, 99.9 atom % d, dichloromethane-d', 99.96 atom % d
dichloro(dideuterio)methane

Specifications

Liquid
Sweet
1665-00-5
MFCD00000882
dichloromethane-d2, methylene chloride-d2, dichloro 2h2 methane, methane-d2, dichloro, cd2cl2, deuterated dichloromethane, dichloro dideuterio methane, dideuteromethylene chloride, dichloromethane-d2, 99.9 atom % d, dichloromethane-d', 99.96 atom % d
YMWUJEATGCHHMB-DICFDUPASA-N
dichloro(dideuterio)methane
160586
99.9%
1.35 g/mL
Moisture
CH2Cl2
UN1593
Insoluble in water. Soluble in acetone,chloroform,tetrachloromethane,dimethylfomrarnide.
C(Cl)Cl
86.939
86.95
Dichloromethane-d2, Isotopic
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H351
Suspected of causing cancer.

P201-P202-P261-P264b-P271-P280i-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P332+P313-P362-P501c

H315-H319-H336-H351

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: DICHLOROMETHANE

missing translation for 'einecsNumber' : 216-776-0

missing translation for 'tsca' : No

Recommended Storage : Ambient

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only