Learn More
Dichloromethylvinylsilane, 97%, Thermo Scientific Chemicals
£14.21 - £246.09
Chemical Identifiers
CAS | 124-70-9 |
---|---|
MDL Number | MFCD00000492 |
InChI Key | YLJJAVFOBDSYAN-UHFFFAOYSA-N |
Synonym | dichloro methyl vinyl silane, methylvinyldichlorosilane, silane, dichloroethenylmethyl, vinylmethyldichlorosilane, methyldichlorovinylsilane, silane, dichloromethylvinyl, unii-b27y6f24py, ccris 2457, dichloromethylvinyl-silan, dichloromethylvinyl silaneShow More |
PubChem CID | 31299 |
IUPAC Name | dichloro-ethenyl-methylsilane |
SMILES | C[Si](C=C)(Cl)Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | |||||
10488100
|
Thermo Scientific Acros
147431000 |
100 g | Glass bottle | N/A | ||||||
10366690
|
Thermo Scientific Acros
147435000 |
500 g | Glass bottle |
£85.63
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10076253
|
Thermo Scientific Acros
147430025 |
2.5 kg | Glass bottle | N/A | ||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
124-70-9 | |
YLJJAVFOBDSYAN-UHFFFAOYSA-N | |
31299 | |
C[Si](C=C)(Cl)Cl |
MFCD00000492 | |
dichloro methyl vinyl silane, methylvinyldichlorosilane, silane, dichloroethenylmethyl, vinylmethyldichlorosilane, methyldichlorovinylsilane, silane, dichloromethylvinyl, unii-b27y6f24py, ccris 2457, dichloromethylvinyl-silan, dichloromethylvinyl silaneShow More | |
dichloro-ethenyl-methylsilane |
Specifications
124-70-9 | |
1.0800g/mL | |
97% | |
H2C=CHSi(CH3)Cl2 | |
MFCD00000492 | |
YLJJAVFOBDSYAN-UHFFFAOYSA-N | |
dichloro-ethenyl-methylsilane | |
31299 | |
≥96.0% | |
Dichloromethylvinylsilane, 97% |
Colorless to Brown | |
Authentic | |
C3H6Cl2Si | |
1.4280 to 1.4310 (20°C, 589nm) | |
dichloro methyl vinyl silane, methylvinyldichlorosilane, silane, dichloroethenylmethyl, vinylmethyldichlorosilane, methyldichlorovinylsilane, silane, dichloromethylvinyl, unii-b27y6f24py, ccris 2457, dichloromethylvinyl-silan, dichloromethylvinyl silane | |
C[Si](C=C)(Cl)Cl | |
141.066 | |
141.07 | |
Liquid |
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Highly flammable liquid and vapor.
Reacts violently with water.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
missing translation for 'einecsNumber' : 204-710-3
missing translation for 'rtecsNumber' : VV3533000
missing translation for 'tsca' : TSCA
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.