missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethyl Ether, Anhydrous, BAKER ANALYZED (ACS), J.T. Baker™
High quality grade chemical that exceeds ACS specifications for laboratory and specialized industrial use. Stabilized with 7ppm BHT.
£131.00 - £402.00
Chemical Identifiers
CAS | 60-29-7 |
---|---|
Molecular Formula | C4H10O |
Molecular Weight (g/mol) | 74.12 |
MDL Number | MFCD00011646 |
InChI Key | RTZKZFJDLAIYFH-UHFFFAOYSA-N |
Synonym | diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether |
PubChem CID | 3283 |
ChEBI | CHEBI:35702 |
IUPAC Name | ethoxyethane |
SMILES | CCOCC |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10443851
|
JT Baker
8254.1000S |
1 L | SAFET |
£131.00
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10231201
|
JT Baker
8254.5000 |
5 L | Aluminum Bottle |
£402.00
5 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Exceeds the requirements set forth by the American Chemical Society (ACS)
- Label provides an actual Lot Analysis as a guarantee of purity and consistency
- Analytical methods used for these determinations are based on standard test procedures described in technical publications
- Improvements developed by J.T. Baker are employed to achieve a higher degree of reliability
Chemical Identifiers
60-29-7 | |
74.12 | |
RTZKZFJDLAIYFH-UHFFFAOYSA-N | |
3283 | |
ethoxyethane |
C4H10O | |
MFCD00011646 | |
diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether | |
CHEBI:35702 | |
CCOCC |
Specifications
-116.3°C | |
Colorless | |
35°C | |
−40°C | |
Sweetish | |
Max. 0.001% | |
Max. 0.0002meq/g | |
MFCD00011646 | |
Max. 0.01% | |
12g/L in water | |
CCOCC | |
74.12 | |
CHEBI:35702 | |
≥99.5% | |
Diethyl ether |
60-29-7 | |
0.71g/cm3 at 20°C | |
1.7 - 48 Vol % | |
170°C | |
Fluid | |
C4H10O | |
(C2H5)2O | |
0.233 mPass at 20°C | |
diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether | |
RTZKZFJDLAIYFH-UHFFFAOYSA-N | |
ethoxyethane | |
3283 | |
74.12g/mol | |
Exceeds ACS specifications |
Safety and Handling
missing translation for 'einecsNumber' : 200-467-2