missing translation for 'onlineSavingsMsg'
Learn More
Diethyl Ether, Extra Pure, SLR, Stabilised with BHT, Fisher Chemical™ Product Code.: 10396080

Diethyl Ether, Extra Pure, SLR, Stabilised with BHT, Fisher Chemical™

Product Code. 10396080
2.5 L, Plastic-coated Glass Bottle
Click to view available options
Quantity:
500 mL
2.5 L
5 L
25 L
Packaging:
Aluminum Can
Metal Can
Plastic-coated Glass Bottle
This item is not returnable. View return policy

Product Code. 10396080

Brand: Fisher Chemical D/2400/PB17

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 60-29-7
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.12
InChI Key RTZKZFJDLAIYFH-UHFFFAOYSA-N
Synonym diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether
PubChem CID 3283
ChEBI CHEBI:35702
IUPAC Name ethoxyethane
SMILES CCOCC

Specifications

Melting Point -116°C
Color Colorless
Boiling Point 34.6°C
Quantity 2.5 L
Packaging Plastic-coated Glass Bottle
Physical Form Liquid
MDL Number MFCD00011646
Vapor Pressure 587mbar at 20°C
Viscosity 0.2448cp at 20°C
Formula Weight 74.12g/mol

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2527135 Certificate TypeCertificate of Analysis Date19/02/2025 Catalog NumberD/2400/MS21,D/2400/27,D/2400/17Z,D/2400/25,D/2400/PB08,D/2400/17,D/2400/PB17
compliance-icons compliance-icons
Product Identifier
  • Diethyl ether
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 1
  • Acute toxicity Category 4
  • Specific target organ toxicity Category 3
Hazard Statement
  • H224-Extremely flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H336-May cause drowsiness or dizziness.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P240-Ground/bond container and receiving equipment.
  • P243-Take precautionary measures against static discharge.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P403+P235-Store in a well-ventilated place. Keep cool.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Diethyl Ether, Extra Pure, SLR, Stabilised with BHT, Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.