missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethyl Ether, Extra Pure, SLR, Stabilised with BHT, Fisher Chemical™
Click to view available options
Quantity:
500 mL
2.5 L
5 L
25 L
Packaging:
Aluminum Can
Metal Can
Plastic-coated Glass Bottle
Specifications
Specifications
Melting Point | -116°C |
Color | Colorless |
Boiling Point | 34.6°C |
Quantity | 500 mL |
Packaging | Plastic-coated Glass Bottle |
Physical Form | Liquid |
MDL Number | 11646 |
Vapor Pressure | 587mbar at 20°C |
Viscosity | 0.2448cp at 20°C |
Formula Weight | 74.12g/mol |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 15
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
1 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
2527135 | Certificate of Analysis | 19/02/2025 | D/2400/MS21,D/2400/27,D/2400/17Z,D/2400/25,D/2400/PB08,D/2400/17,D/2400/PB17 |
Safety and Handling


Product Identifier
- Diethyl ether
- Danger
- Flammable liquid Category 1
- Acute toxicity Category 4
- Specific target organ toxicity Category 3
- H224-Extremely flammable liquid and vapour.
- H302-Harmful if swallowed.
- H336-May cause drowsiness or dizziness.
- EUH066-Repeated exposure may cause skin dryness or cracking.
- EUH019-May form explosive peroxides.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P233-Keep container tightly closed.
- P240-Ground/bond container and receiving equipment.
- P243-Take precautionary measures against static discharge.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P403+P235-Store in a well-ventilated place. Keep cool.
Special Offers
-
Multiple Manufacturers
Chromatography and Chemicals Promotion
Explore our new promotion and get up to 70% off some of our most popular products.Promo Code: 24127Expires: 2025-05-31
New Offer Save Up to 70%
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Diethyl Ether, Extra Pure, SLR, Stabilised with BHT, Fisher Chemical™ >
Spot an opportunity for improvement?Share a Content Correction