missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethyl isobutylmalonate, 98%, Thermo Scientific Chemicals
£24.30 - £222.00
Chemical Identifiers
CAS | 10203-58-4 |
---|---|
Molecular Formula | C11H20O4 |
Molecular Weight (g/mol) | 216.277 |
MDL Number | MFCD00026869 |
InChI Key | OFRFGNSZCYDFOH-UHFFFAOYSA-N |
Synonym | diethyl isobutylmalonate, isobutylmalonic acid diethyl ester, diethyl 2-isobutylmalonate, propanedioic acid, 2-methylpropyl-, diethyl ester, malonic acid, isobutyl-, diethyl ester, diethyl 2-2-methylpropyl propanedioate, ethyl 2-carbethoxy-4-methyl pentanoate, propanedioic acid, 2-2-methylpropyl-, 1,3-diethyl ester, 1,3-diethyl 2-2-methylpropyl propanedioate, diethylisobutylmalonate |
PubChem CID | 25047 |
IUPAC Name | diethyl 2-(2-methylpropyl)propanedioate |
SMILES | CCOC(=O)C(CC(C)C)C(=O)OCC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11307286
|
Thermo Scientific Alfa Aesar
L09202.06 |
5 g |
£24.30
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11317286
|
Thermo Scientific Alfa Aesar
L09202.14 |
25 g |
£76.20
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11327286
|
Thermo Scientific Alfa Aesar
L09202.22 |
100 g | |||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10203-58-4 | |
216.277 | |
OFRFGNSZCYDFOH-UHFFFAOYSA-N | |
25047 | |
CCOC(=O)C(CC(C)C)C(=O)OCC |
C11H20O4 | |
MFCD00026869 | |
diethyl isobutylmalonate, isobutylmalonic acid diethyl ester, diethyl 2-isobutylmalonate, propanedioic acid, 2-methylpropyl-, diethyl ester, malonic acid, isobutyl-, diethyl ester, diethyl 2-2-methylpropyl propanedioate, ethyl 2-carbethoxy-4-methyl pentanoate, propanedioic acid, 2-2-methylpropyl-, 1,3-diethyl ester, 1,3-diethyl 2-2-methylpropyl propanedioate, diethylisobutylmalonate | |
diethyl 2-(2-methylpropyl)propanedioate |
Specifications
10203-58-4 | |
225°C | |
C11H20O4 | |
MFCD00026869 | |
diethyl isobutylmalonate, isobutylmalonic acid diethyl ester, diethyl 2-isobutylmalonate, propanedioic acid, 2-methylpropyl-, diethyl ester, malonic acid, isobutyl-, diethyl ester, diethyl 2-2-methylpropyl propanedioate, ethyl 2-carbethoxy-4-methyl pentanoate, propanedioic acid, 2-2-methylpropyl-, 1,3-diethyl ester, 1,3-diethyl 2-2-methylpropyl propanedioate, diethylisobutylmalonate | |
CCOC(=O)C(CC(C)C)C(=O)OCC | |
216.277 | |
216.28 | |
Diethyl isobutylmalonate |
0.97 | |
>100°C (212°F) | |
1.422 | |
1103030 | |
OFRFGNSZCYDFOH-UHFFFAOYSA-N | |
diethyl 2-(2-methylpropyl)propanedioate | |
25047 | |
98% |
Safety and Handling
missing translation for 'einecsNumber' : 233-504-6
missing translation for 'rtecsNumber' : TY1850200
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only