missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethylene glycol dibutyl ether, 99+%, pure, Thermo Scientific Chemicals
£42.31 - £246.23
Chemical Identifiers
CAS | 112-73-2 |
---|---|
Molecular Formula | C12H26O3 |
Molecular Weight (g/mol) | 218.34 |
InChI Key | KZVBBTZJMSWGTK-UHFFFAOYSA-N |
Synonym | diethylene glycol dibutyl ether, bis 2-butoxyethyl ether, dibutyl carbitol, butyl diglyme, 1-2-2-butoxyethoxy ethoxy butane, 5,8,11-trioxapentadecane, ether, bis butoxyethyl, 2,2'-dibutoxyethyl ether, ether, bis 2-butoxyethyl |
PubChem CID | 8210 |
IUPAC Name | 1-[2-(2-butoxyethoxy)ethoxy]butane |
SMILES | CCCCOCCOCCOCCCC |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10470851
|
Thermo Scientific Acros
194982500 |
250 mL | Glass bottle |
£42.31
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10103880
|
Thermo Scientific Acros
194980010 |
1 L | Glass bottle |
£121.61
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11914741
|
Thermo Scientific Acros
194980025 |
2.5 L | Glass bottle |
£246.23
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
112-73-2 | |
218.34 | |
diethylene glycol dibutyl ether, bis 2-butoxyethyl ether, dibutyl carbitol, butyl diglyme, 1-2-2-butoxyethoxy ethoxy butane, 5,8,11-trioxapentadecane, ether, bis butoxyethyl, 2,2'-dibutoxyethyl ether, ether, bis 2-butoxyethyl | |
1-[2-(2-butoxyethoxy)ethoxy]butane |
C12H26O3 | |
KZVBBTZJMSWGTK-UHFFFAOYSA-N | |
8210 | |
CCCCOCCOCCOCCCC |
Specifications
112-73-2 | |
0.8800g/mL | |
118°C | |
99% min. (GC) | |
[CH3(CH2)3OCH2CH2]2O | |
0.88 | |
Solubility in water: moderate soluble (3g/L at 20°C). Other solubilities: soluble in chloroform | |
CCCCOCCOCCOCCCC | |
218.34 | |
2.39 mPa.s (20°C) | |
99+% | |
Liquid |
-60.0°C | |
256.0°C | |
Authentic | |
C12H26O3 | |
1.4225 to 1.4245 | |
diethylene glycol dibutyl ether, bis 2-butoxyethyl ether, dibutyl carbitol, butyl diglyme, 1-2-2-butoxyethoxy ethoxy butane, 5,8,11-trioxapentadecane, ether, bis butoxyethyl, 2,2'-dibutoxyethyl ether, ether, bis 2-butoxyethyl | |
KZVBBTZJMSWGTK-UHFFFAOYSA-N | |
1-[2-(2-butoxyethoxy)ethoxy]butane | |
8210 | |
218.34 | |
Pure | |
Diethylene glycol dibutyl ether |
Safety and Handling
missing translation for 'einecsNumber' : 204-001-9
RUO – Research Use Only