missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Dehydrated Culture Media: Potato Dextrose Agar

For cultivation of Yeasts and molds. BD Difco™ Dehydrated Culture Media: Potato Dextrose Agar is supplied in dehydrated format.

Brand:  BD 213400

 View more versions of this product

Product Code. 10105824

  • £471.00 / 500g
Estimated Shipment: 13-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Dehydrated format
  • Culture media
  • DNR (Latex) free
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ISO/DIS, 11014, AOAC, BAM, BS12, CCAM, CMPH2, COMPF, EP, JP, MCM9, SMD, USP
Dehydrated
Agar
Isolation of Yeasts and molds
500 g
2°C to 30°C
Potato Dextrose Agar
Potato Starch (from infusion) 4g/L, Dextrose 20g/L, Agar 15g/L
Bottle
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers