Learn More
BD Difco™ Dehydrated Culture Media: Tryptic Soy Agar (Soybean-Casein Digest Agar)
Use dehydrated media for isolation, detection and cultivation of a wide variety of microorganisms. BD Difco™ Dehydrated Culture Media: Tryptic Soy Agar (Soybean-Casein Digest Agar) is supplied in dehydrated form for the convenience of the laboratorian and bioprocessing specialist.
Brand: BD 236930
1102.00 GBP valid until 2024-12-31
Use promo code "21946" to get your promotional price.
Additional Details : Weight : 0.80000kg
Description
Applications:
- Used with or without blood for cultivation and isolation of variety of organisms
- Used for sterility testing
Certifications
AOAC, BAM, CCAM, COMPF, EP, EPA, ISO, JP, SMD, SMWW, USDA, USP
Specifications
AOAC, BAM, CCAM, COMPF, EP, EPA, ISO, JP, SMD, SMWW, USDA, USP | |
Dehydrated | |
Agar | |
Isolation and cultivation of non-fastidious and fastidious microorganisms | |
10 kg |
2°C to 30°C | |
Tryptic Soy Agar | |
Pancreatic digest of Casein 15g/L, Papaic digest of Soybean 5g/L, Sodium Chloride 5g/L, Agar 15g/L | |
Pail |