missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ Dehydrated Culture Media: Tryptic Soy Agar (Soybean-Casein Digest Agar)
Use dehydrated media for isolation, detection and cultivation of a wide variety of microorganisms. BD Difco™ Dehydrated Culture Media: Tryptic Soy Agar (Soybean-Casein Digest Agar) is supplied in dehydrated form for the convenience of the laboratorian and bioprocessing specialist.
£94.80 - £1235.00
Specifications
Certifications/Compliance | AOAC, BAM, CCAM, COMPF, EP, EPA, ISO, JP, SMD, SMWW, USDA, USP |
---|---|
Content And Storage | 2°C to 30°C |
Form | Dehydrated |
Media Type | Tryptic Soy Agar |
Product Type | Agar |
Product Code | Brand | Quantity | Packaging Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging Type | Price | Quantity & Availability | ||||
11768822
|
BD
236950 |
500 g | Bottle |
£94.80
500g |
In Stock
Log in to see stock. |
||||
11748822
|
BD
236920 |
2 kg | Pail |
£489.00
2kg |
Estimated Shipment: 17-12-2024
Log in to see stock. |
||||
11758822
|
BD
236930 |
10 kg | Pail |
£1235.00
10kg |
Estimated Shipment: 22-01-2025
Log in to see stock. |
||||
Description
Applications:
- Used with or without blood for cultivation and isolation of variety of organisms
- Used for sterility testing
Certifications
AOAC, BAM, CCAM, COMPF, EP, EPA, ISO, JP, SMD, SMWW, USDA, USP
Specifications
AOAC, BAM, CCAM, COMPF, EP, EPA, ISO, JP, SMD, SMWW, USDA, USP | |
Dehydrated | |
Agar |
2°C to 30°C | |
Tryptic Soy Agar | |
Pancreatic digest of Casein 15g/L, Papaic digest of Soybean 5g/L, Sodium Chloride 5g/L, Agar 15g/L |
Safety and Handling
missing translation for 'shelfLife' : 1825 Days