Learn More
Digi-Sense™ Borescopes
Cole Parmer Digi-Sense Borescopes features a flexible, camera-tipped probe. Ideal for inspecting duct work, wiring locations, piping and vehicles.
Brand:  Digi-Sense™ 20250-27
 View more versions of this product
Product Code. 15860036
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Includes: Probe, Mirror, Hook, Magnet, Carrying case, USB cable, DC 5V charger, Video output cable, Rechargable 3.7V lithium battery
Description
Description
  • Camera-tipped probe provides easy viewing in hard-to-reach areas
  • Bright LCD monitor allows instant viewing of area to be inspected
  • Adjustable LED light on camera tip provides the necessary light in dimly lit areas or adjusts down when viewing reflective objects to eliminate washout
Compliance
CE; IP67
Services
2 year
Specifications
Specifications
Color LCD
CE; IP67
IP67
4 adjustable LED lights
3.2 in.
Yes
Built-in lithium battery
8 GB flash memory
English, French, German, Spanish
Battery; USB charging
Still and video images for playback
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Digi-Sense™ Borescopes >