Learn More
Diisopropyl sulfide, 99%, Thermo Scientific Chemicals
£40.70 - £122.00
Chemical Identifiers
CAS | 625-80-9 |
---|---|
Molecular Formula | C6H14S |
Molecular Weight (g/mol) | 118.24 |
MDL Number | MFCD00008895 |
InChI Key | XYWDPYKBIRQXQS-UHFFFAOYSA-N |
Synonym | diisopropyl sulfide, isopropyl sulfide, propane, 2,2'-thiobis, 2,4-dimethyl-3-thiapentane, isopropyl sulphide, diisopropyl sulphide, 2,2'-thiodipropane, 2-isopropylsulfanyl propane, 2-propan-2-ylsulfanyl propane, isopropylsulfide |
PubChem CID | 12264 |
SMILES | CC(C)SC(C)C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11499207
|
Thermo Scientific Alfa Aesar
B21302.09 |
10 g |
£40.70
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11409217
|
Thermo Scientific Alfa Aesar
B21302.18 |
50 g |
£122.00
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Used as a flavoring agents and intermediate for organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsUsed as a flavoring agents and intermediate for organic synthesis.
Solubility
Not miscible or difficult to mix with water.
Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Chemical Identifiers
625-80-9 | |
118.24 | |
XYWDPYKBIRQXQS-UHFFFAOYSA-N | |
12264 |
C6H14S | |
MFCD00008895 | |
diisopropyl sulfide, isopropyl sulfide, propane, 2,2'-thiobis, 2,4-dimethyl-3-thiapentane, isopropyl sulphide, diisopropyl sulphide, 2,2'-thiodipropane, 2-isopropylsulfanyl propane, 2-propan-2-ylsulfanyl propane, isopropylsulfide | |
CC(C)SC(C)C |
Specifications
625-80-9 | |
119°C to 120°C | |
Stench | |
(CH3)2CHSCH(CH3)2 | |
MFCD00008895 | |
1731257 | |
Not miscible or difficult to mix with water. | |
CC(C)SC(C)C | |
118.24 | |
118.24 | |
Diisopropyl sulfide |
0.814 | |
7°C (44°F) | |
C6H14S | |
1.4385 | |
UN1993 | |
diisopropyl sulfide, isopropyl sulfide, propane, 2,2'-thiobis, 2,4-dimethyl-3-thiapentane, isopropyl sulphide, diisopropyl sulphide, 2,2'-thiodipropane, 2-isopropylsulfanyl propane, 2-propan-2-ylsulfanyl propane, isopropylsulfide | |
XYWDPYKBIRQXQS-UHFFFAOYSA-N | |
2-propan-2-ylsulfanylpropane | |
12264 | |
99% |
Safety and Handling
GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H225-H315-H319-H335
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
missing translation for 'einecsNumber' : 210-911-7
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only