Learn More
Day Impex™ Dilvac™ Cylindrical Glass Dewar Flask
Standard silvered glass dewar skillfully constructed of borosilicate glass. Day Impex™ Dilvac™ Cylindrical Glass Dewar Flask is ultra-low evacuated for safe and efficient handling of liquified gases, freezing mixtures, and specimens.
Brand: Day Impex™ 210024
Code : 03
Additional Details : Weight : 0.01000kg
Description
- Wide range of sizes and capacities
- Borosilicate glass
- Ultra evacuated
- Silvered
Specifications
50 mm | |
232mm | |
200mm | |
375 mL |
Liquified gases, freezing mixtures, and specimens. | |
Borosilicate Glass | |
67mm |