Learn More
Dimethoxymethane, 99.5+%, Thermo Scientific Chemicals
£42.90 - £412.00
Chemical Identifiers
CAS | 109-87-5 |
---|---|
Molecular Formula | C3H8O2 |
Molecular Weight (g/mol) | 76.09 |
MDL Number | MFCD00008495 |
InChI Key | NKDDWNXOKDWJAK-UHFFFAOYSA-N |
Synonym | methylal, formal, methane, dimethoxy, formaldehyde dimethyl acetal, dimethyl formal, anesthenyl, 2,4-dioxapentane, bis methoxy methane, methylene dimethyl ether, methoxymethyl methyl ether |
PubChem CID | 8020 |
ChEBI | CHEBI:48341 |
IUPAC Name | dimethoxymethane |
SMILES | COCOC |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10498250
|
Thermo Scientific Acros
115560010 |
1 L | Glass bottle |
£42.90
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10264570
|
Thermo Scientific Acros
115560025 |
2.5 L | Glass bottle |
£67.00
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10073563
|
Thermo Scientific Acros
115560250 |
25 L | Metal drum |
£412.00
25 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
109-87-5 | |
76.09 | |
NKDDWNXOKDWJAK-UHFFFAOYSA-N | |
8020 | |
dimethoxymethane |
C3H8O2 | |
MFCD00008495 | |
methylal, formal, methane, dimethoxy, formaldehyde dimethyl acetal, dimethyl formal, anesthenyl, 2,4-dioxapentane, bis methoxy methane, methylene dimethyl ether, methoxymethyl methyl ether | |
CHEBI:48341 | |
COCOC |
Specifications
109-87-5 | |
Colorless | |
41.0°C to 42.0°C | |
Authentic | |
C3H8O2 | |
1.3525 to 1.3545 | |
01,671; 02,271; 06,374; 07,115 | |
0.86 | |
Solubility in water: 32.3g/100 mL (16°C). Other solubilities: miscible with alcohol,ether,oils,hydrocarbons,miscible with most common organic solvents,very soluble in acetone,benzene | |
COCOC | |
76.09 | |
CHEBI:48341 | |
76.09 | |
Liquid |
-105.0°C | |
0.8600g/mL | |
−18°C | |
99.5% min. (GC) | |
CH2(OCH3)2 | |
MFCD00008495 | |
15, 6086 | |
methylal, formal, methane, dimethoxy, formaldehyde dimethyl acetal, dimethyl formal, anesthenyl, 2,4-dioxapentane, bis methoxy methane, methylene dimethyl ether, methoxymethyl methyl ether | |
NKDDWNXOKDWJAK-UHFFFAOYSA-N | |
dimethoxymethane | |
8020 | |
3.25 mPa.s (20°C) | |
99.5+% | |
Dimethoxymethane |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapor.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Ground/Bond container and receiving equipment.
Store in a well-ventilated place.
Keep container tightly closed.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 203-714-2
- Dimethoxymethane
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
Hazard Statement
- H225-Highly flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
RUO – Research Use Only