missing translation for 'onlineSavingsMsg'
Learn More
Dimethyl Sulfoxide, ≥99% (Molecular Biology/ACS Certified), MP Biomedicals Product Code.: 11440522
compliance-icons

Dimethyl Sulfoxide, ≥99% (Molecular Biology/ACS Certified), MP Biomedicals

Product Code. 11440522 Shop All MP Biomedicals Products
100 mL
Click to view available options
Quantity:
100 mL
1 L
This item is not returnable. View return policy

Product Code. 11440522

Brand: MP Biomedicals 0219141880

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 67-68-5
Molecular Formula C2H6OS
Molecular Weight (g/mol) 78.13
MDL Number MFCD00002089
InChI Key IAZDPXIOMUYVGZ-UHFFFAOYSA-N
Synonym dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexideShow MoreShow Less
PubChem CID 679
ChEBI CHEBI:28262
SMILES CS(C)=O

Specifications

Quantity 100 mL
IUPAC Name methanesulfinylmethane
Formula Weight 78.13
Grade Certified ACS Reagent
Chemical Name or Material Dimethyl Sulfoxide
compliance-icons
Product Identifier
  • dimethylsulphoxide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P362-Take off contaminated clothing.
Supplemental information
  • MIXTURE LIST-Contains : dimethyl sulphoxide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Dimethyl Sulfoxide, ≥99% (Molecular Biology/ACS Certified), MP Biomedicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.