missing translation for 'onlineSavingsMsg'
Learn More

DimethylFormamide, Certified AR for Analysis, Fisher Chemical™

£83.00 - £1170.00

Chemical Identifiers

CAS 68-12-2
Molecular Formula C3H7NO
Molecular Weight (g/mol) 73.10
MDL Number MFCD00003284
InChI Key ZMXDDKWLCZADIW-UHFFFAOYSA-N
Synonym dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfa
PubChem CID 6228
ChEBI CHEBI:17741
IUPAC Name N,N-dimethylformamide
SMILES CN(C)C=O
View More Specs

Products 7
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10191560
View Documents
Fisher Chemical
D/3841/08
500 mL Amber glass bottle
£83.00
500mL
Please to purchase this item. Need a web account? Register with us today!
10560911
View Documents
Fisher Chemical
D/3841/15
1 L Amber glass bottle
£134.00
1 L
Please to purchase this item. Need a web account? Register with us today!
10284140
View Documents Promotion Details
Fisher Chemical
D/3841/17
2.5 L Amber glass bottle
£225.00
2.50 L
Please to purchase this item. Need a web account? Register with us today!
10273230
View Documents
Fisher Chemical
D/3841/PB17
2.5 L Plastic Coated Glass bottle
£267.00
2.50 L
Please to purchase this item. Need a web account? Register with us today!
11483883
View Documents
Fisher Chemical
D/3841/21
5 L HDPE plastic bottle
£373.00
5 L
Please to purchase this item. Need a web account? Register with us today!
12633356
View Documents
Fisher Chemical
D/3841/24
10 L Plastic container
£490.00
10 L
Please to purchase this item. Need a web account? Register with us today!
10294140
View Documents
Fisher Chemical
D/3841/25
25 L Plastic container
£1170.00
25 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

DimethylFormamide, Certified AR for Analysis

Specifications

Chemical Identifiers

68-12-2
73.10
ZMXDDKWLCZADIW-UHFFFAOYSA-N
6228
N,N-dimethylformamide
C3H7NO
MFCD00003284
dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfa
CHEBI:17741
CN(C)C=O

Specifications

68-12-2
Colorless
C3H7NO
dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfa
CN(C)C=O
73.10
CHEBI:17741
4.9mbar at 20°C
-61°C
153°C
MFCD00003284
ZMXDDKWLCZADIW-UHFFFAOYSA-N
N,N-dimethylformamide
6228
73.09g/mol
Liquid
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Dimethylformamide

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 1B

Hazard Statement
  • H226-Flammable liquid and vapour.
  • H312-Harmful in contact with skin.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H360D-May damage the unborn child.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.