missing translation for 'onlineSavingsMsg'
Learn More
Dimethylglyoxime, disodium salt octahydrate, 99%, Thermo Scientific Chemicals Product Code.: 10209700

Dimethylglyoxime, disodium salt octahydrate, 99%, Thermo Scientific Chemicals

Product Code. 10209700
100 g, Glass bottle
Click to view available options
Quantity:
100 g
250 g
1 kg
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10209700

Brand: Thermo Scientific Acros 408341000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Dimethylglyoxime, disodium salt octahydrate, 99%, C4H6N2Na2O2.8H2O, CAS Number-75006-64-3, c4h6n2o2.2na.8h2o, dimethylglyoxime di-sodium salt octahydrate, dimethylglyoxime disodium salt octahydrate, sodium dimethylglyoxime octahydrate, 1kg, White to Beige, 262-523-2, 304.19

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75006-64-3
Molecular Formula C4H22N2Na2O10
Molecular Weight (g/mol) 304.20
MDL Number MFCD00150288
InChI Key IJJDRGUPRATWQX-UHFFFAOYSA-L
Synonym dimethylglyoxime disodium salt octahydrate, c4h6n2o2.2na.8h2o, sodium dimethylglyoxime octahydrate, dimethylglyoxime di-sodium salt octahydrateShow MoreShow Less
PubChem CID 71310106
SMILES O.O.O.O.O.O.O.O.[Na+].[Na+].CC(=N[O-])C(C)=N[O-]

Specifications

Melting Point >300.0°C
Color White to Beige
Loss on Drying 46% to 49%
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula CH3C(NONa)C(NONa)CH3·8H2O
Quantity 100 g
Solubility Information (50mg/ml,50% ethanol) Clear solution
IUPAC Name disodium;2-N,3-N-dioxidobutane-2,3-diimine;octahydrate
Formula Weight 304.19
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Dimethylglyoxime, disodium salt octahydrate, 99%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0466814 Certificate TypeCertificate of Analysis Date18/10/2024 Catalog Number408342500,408340010,408341000
Lot NumberA0464545 Certificate TypeCertificate of Analysis Date12/07/2024 Catalog Number408342500,408340010,408341000
Lot NumberA0461195 Certificate TypeCertificate of Analysis Date21/03/2024 Catalog Number408342500,408340010,408341000
Lot NumberA0457426 Certificate TypeCertificate of Analysis Date03/11/2023 Catalog Number408342500,408340010,408341000
Lot NumberA0449947 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number408342500,408340010,408341000
Product Identifier
  • Dimethylglyoxime, disodium salt octahydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Dimethylglyoxime, disodium salt octahydrate, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.