missing translation for 'onlineSavingsMsg'
Learn More
Dioctyl sulfosuccinate sodium salt, 96%, Thermo Scientific Chemicals Product Code.: 11329708

Dioctyl sulfosuccinate sodium salt, 96%, Thermo Scientific Chemicals

Product Code. 11329708
500 g
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy

Product Code. 11329708

Brand: Thermo Scientific Alfa Aesar 044203.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Wetting and solubilizing agent

Wetting and solubilizing agent.Dioctyl sulfosuccinate sodium salt is used as a surfactant, wetting agent and in the preparation of reverse micelles. It is utilized in electrophoresis detection of natural and synthetic estrogens. It also finds use in the drug, cosmetics, food industry and as a laxative to treat constipation. Further, it is used as a food additive, emulsifier and dispersant. It plays an important role as an excipient in the production of tablets and suspensions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 577-11-7
Molecular Formula C20H37NaO7S
Molecular Weight (g/mol) 444.56
MDL Number MFCD00012455
InChI Key APSBXTVYXVQYAB-UHFFFAOYNA-M
Synonym docusate sodium, dioctyl sodium sulfosuccinate, aerosol ot, constonate, diox, manoxol ot, diomedicone, clestol, complemix, defilin
PubChem CID 23673837
IUPAC Name sodium;1,4-bis(2-ethylhexoxy)-1,4-dioxobutane-2-sulfonate
SMILES [Na+].CCCCC(CC)COC(=O)CC(C(=O)OCC(CC)CCCC)S([O-])(=O)=O

Specifications

Melting Point 153°C to 157°C
Density 1.1 g/mL
Quantity 500 g
Sensitivity Hygroscopic
Merck Index 14,3401
Solubility Information Soluble in water and methanol.
Formula Weight 444.55
Percent Purity 96%
Physical Form Solid
Chemical Name or Material Dioctyl sulfosuccinate sodium salt

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberX21K006 Certificate TypeCertificate of Analysis Date05/11/2024 Catalog Number044203.36,044203.22
Lot NumberT30K015 Certificate TypeCertificate of Analysis Date13/08/2024 Catalog Number044203.36,044203.22
Lot NumberP19K018 Certificate TypeCertificate of Analysis Date02/04/2024 Catalog Number044203.36,044203.22
Lot NumberN22I012 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number044203.36,044203.22
Lot NumberN12H020 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number044203.36,044203.22
compliance-icons
Product Identifier
  • Dioctyl sulfosuccinate sodium salt
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Dioctyl sulfosuccinate sodium salt

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Dioctyl sulfosuccinate sodium salt, 96%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.