missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Dionex™ Fused Silica Tubing

Thermo Scientific™ Fused Silica Tubing provides excellent chemical durability and inertness.

Brand:  Thermo Scientific™ 160477

 View more versions of this product

Product Code. 11342123

  • £219.09 / Each
Estimated Shipment: 05-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Thermo Scientific™ Fused Silica Tubing provides excellent chemical durability and inertness.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

50 um I.D.; 280 um O.D.; 5 m length
Fused Silica
Capillary Tubing
50 μm
LC-Packings Ultimate Capillary/Nano HPLC
nanoLC
5 m
280 μm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers