Learn More
Thermo Scientific™ Dionex™ Fused Silica Tubing
Thermo Scientific™ Fused Silica Tubing provides excellent chemical durability and inertness.
Brand:  Thermo Scientific™ 160480
 View more versions of this product
Product Code. 11372123
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Thermo Scientific™ Fused Silica Tubing provides excellent chemical durability and inertness.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
150 um I.D.; 280 um O.D.; 5 m length
Fused Silica
Capillary Tubing
150 μm
LC-Packings Ultimate Capillary/Nano HPLC
nanoLC
5 m
280 μm