Learn More
Thermo Scientific™ Dionex™ Specially Formulated PTFE Tubing
Thermo Scientific™ Dionex Specially Formulated PTFE Tubing is used as connector for fused silica tubing (nano and cap LC). Choose fitting dimensions for OD of fused silica tubing used.
Brand: Thermo Scientific™ 161030
Code : 16
Additional Details : Weight : 0.10000kg
Description
Thermo Scientific™ Dionex Specially Formulated PTFE Tubing is used as connector for fused silica tubing (nano and cap LC).
Specifications
350 um ID; Low pressure | |
Fused Silica Capillary Tubing | |
nanoLC | |
Capillary Tubing |