missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ Dip Sampler

A dip sampler for the simple removal of liquid samples, with a good view provided by transparent tubes.

Brand:  Buerkle™ 5315-0200

63.99 GBP valid until 2024-12-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 10036611

  • £120.00 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Upper end with reflux valve, lower end with ball valve
  • Manufactured from transparent polypropylene
  • All-layer and point sampling possible
Specifications

Specifications

Dip Sampler
Plunging siphon
Transparent
25 mm
Polypropylene
500 mL
For Simple Removal of Liquid Samples, Multi-Layer and Point Sampling
Immersion depth of 100 to 200 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.