missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dipentyl phthalate, 97%, Thermo Scientific Chemicals
£120.60 - £1254.24
Chemical Identifiers
CAS | 131-18-0 |
---|---|
Molecular Formula | C18H26O4 |
Molecular Weight (g/mol) | 306.4 |
InChI Key | IPKKHRVROFYTEK-UHFFFAOYSA-N |
Synonym | dipentyl phthalate, di-n-pentyl phthalate, diamyl phthalate, amyl phthalate, amoil, di-n-amyl phthalate, di-n-pentylphthalate, phthalic acid, dipentyl ester, phthalic acid diamyl ester, 1,2-benzenedicarboxylic acid, dipentyl ester |
PubChem CID | 8561 |
ChEBI | CHEBI:34680 |
IUPAC Name | dipentyl benzene-1,2-dicarboxylate |
SMILES | CCCCCOC(=O)C1=CC=CC=C1C(=O)OCCCCC |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10778352
|
Thermo Scientific Acros
357360250 |
25 g | Glass bottle |
£120.60
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10276051
|
Thermo Scientific Acros
357361000 |
100 g | Glass bottle |
£327.63
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11955041
|
Thermo Scientific Acros
357365001 |
500 g | Glass bottle |
£1254.24
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
131-18-0 | |
306.4 | |
dipentyl phthalate, di-n-pentyl phthalate, diamyl phthalate, amyl phthalate, amoil, di-n-amyl phthalate, di-n-pentylphthalate, phthalic acid, dipentyl ester, phthalic acid diamyl ester, 1,2-benzenedicarboxylic acid, dipentyl ester | |
CHEBI:34680 | |
CCCCCOC(=O)C1=CC=CC=C1C(=O)OCCCCC |
C18H26O4 | |
IPKKHRVROFYTEK-UHFFFAOYSA-N | |
8561 | |
dipentyl benzene-1,2-dicarboxylate |
Specifications
131-18-0 | |
Colorless to Yellow | |
342.0°C | |
96% min. (GC) | |
(C(O)OC5H11)2C6H4 | |
1.023 | |
IPKKHRVROFYTEK-UHFFFAOYSA-N | |
dipentyl benzene-1,2-dicarboxylate | |
8561 | |
306.4 | |
Liquid |
-55.0°C | |
1.0230g/mL | |
Authentic | |
C18H26O4 | |
1.4885 to 1.4905 | |
dipentyl phthalate, di-n-pentyl phthalate, diamyl phthalate, amyl phthalate, amoil, di-n-amyl phthalate, di-n-pentylphthalate, phthalic acid, dipentyl ester, phthalic acid diamyl ester, 1,2-benzenedicarboxylic acid, dipentyl ester | |
CCCCCOC(=O)C1=CC=CC=C1C(=O)OCCCCC | |
306.4 | |
CHEBI:34680 | |
97% | |
Dipentyl phthalate |
Safety and Handling
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 205-017-9
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Dipentyl phthalate
Signal Word
- Danger
Hazard Category
- ACUTE AQUATIC Acute 1
- Reproductive toxicity Category 1B
Hazard Statement
- H400-Very toxic to aquatic life.
- H360FD-May damage fertility. May damage the unborn child.
Precautionary Statement
- P201-Obtain special instructions before use.
- P273-Avoid release to the environment.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title