missing translation for 'onlineSavingsMsg'
Learn More
Diphenyleneiodonium chloride, Thermo Scientific Chemicals Product Code.: 15426609

Diphenyleneiodonium chloride, Thermo Scientific Chemicals

Product Code. 15426609
50 mg
Click to view available options
Quantity:
50 mg
100 mg
This item is not returnable. View return policy

Product Code. 15426609

Brand: Thermo Scientific Alfa Aesar J64838.MF

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4673-26-1
Molecular Formula C12H8ClI
Molecular Weight (g/mol) 314.55
MDL Number MFCD00214165
InChI Key FCFZKAVCDNTYID-UHFFFAOYSA-M
Synonym diphenyleneiodonium chloride, dibenziodolium chloride, unii-7m9d81yz2n, dpi, 8? 3-iodatricyclo 7.4.0.0 2 ,? trideca-1 13 ,2,4,6,9,11-hexaen-8-ylium chloride, dibenziodolium, chloride, d0n0gj, c12h8i.cl, dibenzo b,d iodolium chloride, 2,2'-biphenylyleneiodonium chlorideShow MoreShow Less
PubChem CID 2733504
ChEBI CHEBI:77967
SMILES C1=CC=C2C(=C1)C3=CC=CC=C3[I+]2.[Cl-]

Specifications

Melting Point >310°C
Color White
Quantity 50 mg
Formula Weight 314.55
Physical Form Powder
Chemical Name or Material Diphenyleneiodonium chloride

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberM26K021 Certificate TypeCertificate of Analysis Date09/02/2024 Catalog NumberJ64838.MF,J64838.MC
Lot NumberN11I026 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberJ64838.MF,J64838.MC
Lot NumberT09B058 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberJ64838.MF,J64838.MC
Lot NumberR28F011 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberJ64838.MF,J64838.MC
Lot NumberW27H005 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberJ64838.MF,J64838.MC
compliance-icons
Product Identifier
  • Diphenyleneiodonium chloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Diphenyleneiodoniumchloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Diphenyleneiodonium chloride, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.