Learn More
Thermo Scientific™ Replacement Tips for UNIFILTER and UNIGUARD Direct-Connection Holders
Standard replacement tips for Thermo Scientific™ UNIGUARD™ and UNIFILTER™ Direct-Connection Holders.
Brand: Thermo Scientific™ 850-RT
Code : 16
Additional Details : Weight : 0.10000kg
Description
Use the Thermo Scientific™ Replacement Tip as a standard component for UNIFILTER™ Direct-Connection HPLC Filter Systems or for UNIGUARD™ Direct-Connection Guard Cartridge Holders. The UNIGUARD Cartridge Holder connectors have an inner diameter that matches the analytical column inset and eliminates the need for extra fittings. The UNIFILTER Filter Systems are replaceable 0.5μm drop-in filters that enhance column lifetime and improve performance. A Filter System comprises the holder, filter and replacement tip.
Specifications
Standard Replacement Tip, CPI | |
UNIFILTER HPLC Filter Systems and UNIGUARD Guard Cartridge Holders |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.