missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ Direct Overhead Drive
For 8 L, 15 L, or 36 L paddle assemblies.
Brand: Corning™ 400649
This item is not returnable.
View return policy
Description
- For 8 L, 15 L, or 36 L paddle assemblies.
- Used on all series 4510 and 4512 Spinner flasks.
Specifications
120 mm | |
Proculture Spinner Flasks |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction