missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Direct Overhead Drive

For 8 L, 15 L, or 36 L paddle assemblies.

£2914.57

Specifications

For Use With (Equipment) Proculture Spinner Flasks
View More Specs

Products 2
Product Code Brand Diameter (Metric) Center Neck Price Quantity & Availability  
Product Code Brand Diameter (Metric) Center Neck Price Quantity & Availability  
11333755
View Documents
Corning™
400649
120 mm
£2914.57
Each
Estimated Shipment: 17-07-2025
to see stock.
Add to basket
 
11972011
View Documents
Corning™
402614
100 mm
£2914.57
Each
Estimated Shipment: 17-07-2025
to see stock.
Add to basket
 
Description

Description

  • For 8 L, 15 L, or 36 L paddle assemblies.
  • Used on all series 4510 and 4512 Spinner flasks.
Specifications

Specifications

Proculture Spinner Flasks
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Corning™ Direct Overhead Drive

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.