Learn More
Veolia Water Systems™ Disinfection Tablets
Use Disinfection Tablets to sanitise tubing, or pipework and system parts that come into contact with water, to remove bacteria, reduce bio film and protect the system against bacterial growth.
Brand:  Veolia Water Systems™ CT3
Product Code. 12826753
  • £133.00 / Pack of 12
Explore more special offers
Description
Description
  • For regular disinfection of system parts that come into contact with water
Specifications
Specifications
Effervescent tablets
Disinfection Tablets
Sanitising Tubing and Pipework
0.99 kg
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Veolia Water Systems™ Disinfection Tablets > Product Type: Disinfection Tablets