Learn More
Epredia™ Disposable Base Molds
Use Epredia™ Disposable Base Molds with all standard-size cassettes.
Brand:  Epredia™ 58952
111.35 GBP valid until 2024-12-31
Use promo code "21628" to get your promotional price.
 View more versions of this product
Product Code. 12510117
  • £145.00 / Pack of 500
Explore more special offers
Description
Description
  • Cleaning associated with metal molds is not required
  • Transparency facilitates tissue orientation
Ordering Information:
  • Sold in cases of four boxes.
Specifications
Specifications
Base Mold
Plastic
30 x 24 mm
Single-use