missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ Disposable Particulate Respirator, 8000 Series
Ensures comfortable wear over a range of face sizes. The 3M 8000 Series Particulate Respirator has a convex shape.
Brand: 3M™ 7000034734
This item is not returnable.
View return policy
Description
Ensures comfortable wear over a range of face sizes. The 3M 8000 Series Particulate Respirator has a convex shape.
The 3M™ Respirators 8000 Series provide lightweight, comfortable and effective respiratory protection against dust and mist. The convex shape, twin strap design, nose foam and nose clip ensure comfortable wear over a range of face sizes. The collapse resistant shell and proprietary 3M™ respirators offer durable, comfortable protection particularly in hot and humid conditions. Dusts, mists and metal fumes are the most common respiratory hazards in the working environment as by-products of processes such as grinding and mixing. 3M respirators provide comfortable, maintenance-free protection against these particular hazards.
Specifications
European Community Directive 89/686/EEC, EN 149:2001 A1:2009, CE | |
Single-use | |
General Purpose Particulate Respirator | |
FFP1 NR D | |
Yes | |
20/Pk. | |
Convex | |
No |
Cup/Molded | |
FFP1 NR D | |
No | |
Protection against dust and mist | |
8710 | |
Dusts, mists and Metal fumes | |
Non-sterile |
Product Suggestions
Customers who viewed this item also viewed
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
3M™ Disposable Particulate Respirator, 8000 Series > Filter Type: FFP1 NR D; With valve; Qty: 20 Pack
Spot an opportunity for improvement?Share a Content Correction