missing translation for 'onlineSavingsMsg'
Learn More

3M™ Disposable Particulate Respirator, 8000 Series

Ensures comfortable wear over a range of face sizes. The 3M 8000 Series Particulate Respirator has a convex shape.

Brand:  3M™ 7100101091

15.37 GBP valid until 2025-04-30
Use promo code "24155" to get your promotional price.


 View more versions of this product

Product Code. 11733363

  • £31.60 / Pack of 10
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

Ensures comfortable wear over a range of face sizes. The 3M 8000 Series Particulate Respirator has a convex shape.

The 3M™ Respirators 8000 Series provide lightweight, comfortable and effective respiratory protection against dust and mist. The convex shape, twin strap design, nose foam and nose clip ensure comfortable wear over a range of face sizes. The collapse resistant shell and proprietary 3M™ respirators offer durable, comfortable protection particularly in hot and humid conditions. Dusts, mists and metal fumes are the most common respiratory hazards in the working environment as by-products of processes such as grinding and mixing. 3M respirators provide comfortable, maintenance-free protection against these particular hazards.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

EN 149
Single-use
General Purpose Particulate Respirator
FFP2
10/Pk.
Hull
No
Cup/Molded
FFP2
Yes
For dusts, Fibers and aqueous mists
For dusts, Fibers and aqueous mists
Non-sterile
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales

  • Multiple Manufacturers
    Save up to 60% on gloves, apparel, and more
    Promo Code: 24131

    Expires2025-06-08

    New Offer
    Shop Now
Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto
3M™ Disposable Particulate Respirator, 8000 Series > Filter Type: FFP2; Qty: 10 Pack

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado