Learn More
Kartell™ Technicon™ Beckman™, Konelab™ Polystyrene Sample Cups
Manufactured with crystal clear polystyrene for optimum visibility. Kartell™ Technicon™ Beckman™ Konelab™ Sample Cups are used for specimen analysis.
£89.20 - £213.00
Specifications
Material Polystyrene
Disposable Single-use
View More Specs
Products 3
Product Code Brand Capacity (Metric) Height (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Height (Metric) Price Quantity & Availability  
12039877
View Documents
Kartell™
02508004
4 mL 37.9 mm
£213.00
Pack of 1000
Estimated Shipment: 02-06-2025
to see stock.
Add to basket
11303814
View accessory products View Documents
Kartell™
02502004
2 mL 24.9 mm
£112.44
Pack of 1000
In Stock
to see stock.
Edge
Add to basket
12523327
View accessory products View Documents
Kartell™
02501004
1.5 mL 22.6
£89.20
Pack of 1000
Estimated Shipment: 05-06-2025
to see stock.
Add to basket
Description
Description
Manufactured with crystal clear polystyrene for optimum visibility. Kartell™ Technicon™ Beckman™ Konelab™ Sample Cups are used for specimen analysis.
  • A range of crystal clear polystyrene auto analyser cups with conical bases
  • For Technicon™, Beckman™, Konelab™
  • Supplied without caps
  • Graduated
Specifications
Specifications
Polystyrene
Single-use
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Kartell™ Technicon™ Beckman™, Konelab™ Polystyrene Sample Cups