missing translation for 'onlineSavingsMsg'
Learn More
Learn More
New Source™ Disposable Chemical Resistant SMS Coveralls Type 5/6
Disposable chemical resistant coveralls offering protection against solid particulates and liquid splashes and sprays. CE certified as PPE Cat III under Regulation EU 2016/425
£6.25 - £25.00
Specifications
Material | Non-woven SMS |
---|---|
Disposable | Single-use |
Ankle Style | Elastic |
Cuff Style | Elastic |
Closure Type | Flap |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
16691793
|
New Source™
HY9766M |
Medium | N/A | ||||||
16645042
|
New Source™
HY9766L |
Large | N/A | ||||||
16655042
|
New Source™
HY9766XL |
X-Large | N/A | ||||||
16665042
|
New Source™
HY9766XXL |
2X-Large | N/A | ||||||
17004287
|
New Source™
HY9766XXXL |
3X-Large | N/A | ||||||
Description
- Conforms to CE Category III Type 5/6
- Non-woven SMS disposable fabric
- Protects against chemical and biological hazards
Specifications
Non-woven SMS | |
Elastic | |
Flap | |
55 gsm |
Single-use | |
Elastic | |
CE Cat III PPE meeting: EN ISO 13982-1:2004, EN ISO 13034: 2005 +A1:2009, EN ISO 14325:2004 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title