missing translation for 'onlineSavingsMsg'
Learn More

Lenz Laborglasinstrumente™ Distilling Links

Distilling Links for the connection of distillation thermometers of 55 mm built-in length.

Brand:  Lenz Laborglasinstrumente™ 05053201

 View more versions of this product

Product Code. 10702682

  • £69.70 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • With Liebig condenser
  • Outlet with NS cone
  • Water connection with glass olives, with NS socket
Specifications

Specifications

For distillation thermometers of 55 mm built-In length
Duran Glass
160 mm
Distilling Link
NS 14/23, NS 14/23
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers