missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lenz Laborglasinstrumente™ Distilling Links with Liebig Condenser and Head to Claisen
Distilling Links with Liebig Condenser and Head to Claisen made of DURAN™ tubing.
Brand: Lenz Laborglasinstrumente™ 05057304
This item is not returnable.
View return policy
Specifications
For distillation thermometers of 55 mm built-In length | |
Duran Glass | |
250 mm |
Distilling Link | |
NS 14/23, NS 29/32 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction