Learn More
Lenz Laborglasintrumente Distribution Tubes With Frit
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand:  Lenz Laborglasintrumente 05432204
 View more versions of this product
Product Code. 10336071
  • £91.90 / Each
Estimated Shipment: 14-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Distribution tubes with frit (porosity: 0)
  • For specific light solvents
  • Made of DURAN™ tubing
Specifications
Specifications
500 mL
Specific light solvents