Learn More
Thermo Scientific Chemicals DL-1,4-Dithiothreitol, 99%, for biochemistry
1,4-Dithiothreitol, CAS # 3483-12-3, is a reducing agent commonly used to reduce disulphide bonds in protein studies, such as western blot SDS-page electrophoresis.
£42.11 - £4833.05
Chemical Identifiers
CAS | 3483-12-03 |
---|---|
MDL Number | MFCD00004877 |
InChI Key | VHJLVAABSRFDPM-IMJSIDKUSA-N |
Synonym | dithiothreitol, dl-1,4-dithiothreitol, dl-dithiothreitol, 1,4-dithio-dl-threitol, d-1,4-dithiothreitol, d-dtt, 2s,3s-1,4-dimercaptobutane-2,3-diol, threo-1,4-dimercapto-2,3-butanediol, 1,4-dithiothreitol, dtt |
PubChem CID | 446094 |
ChEBI | CHEBI:42170 |
IUPAC Name | (2S,3S)-1,4-bis(sulfanyl)butane-2,3-diol |
SMILES | C(C(C(CS)O)O)S |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10448730
|
Thermo Scientific Chemicals
165680010 |
1 g |
£42.11
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10458730
|
Thermo Scientific Chemicals
165680050 |
5 g |
£134.67
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10215550
|
Thermo Scientific Chemicals
165680250 |
25 g |
£431.15
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10063023
|
Thermo Scientific Chemicals
165681000 |
100 g |
£1370.82
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15274654
|
Thermo Scientific Chemicals
165685000 |
500 g |
£4833.05
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General description
- 1,4-Dithiothreitol (DTT or Cleland's reagent) reduces disulfide bonds in proteins, leading to loss of native conformation
- DTT is not stable in solution and should be prepared in small aliquots prior to use; the use of EDTA in the solution can increase the half-life of DTT
- Typically used at a final concentration of 0.1M for protein stabilization; recommended to create a 1M stock
Applications
- Protects thiol groups from oxidation and acts as a chemical chelator in solution; maintains proteins in the denatured state
- Common applications include SDS-page electrophoresis and protein isolation
Chemical Identifiers
3483-12-03 | |
VHJLVAABSRFDPM-IMJSIDKUSA-N | |
446094 | |
(2S,3S)-1,4-bis(sulfanyl)butane-2,3-diol |
MFCD00004877 | |
dithiothreitol, dl-1,4-dithiothreitol, dl-dithiothreitol, 1,4-dithio-dl-threitol, d-1,4-dithiothreitol, d-dtt, 2s,3s-1,4-dimercaptobutane-2,3-diol, threo-1,4-dimercapto-2,3-butanediol, 1,4-dithiothreitol, dtt | |
CHEBI:42170 | |
C(C(C(CS)O)O)S |
Specifications
3483-12-03 | |
39°C to 44°C | |
Authentic | |
C4H10O2S2 | |
MFCD00004877 | |
15, 3419 | |
VHJLVAABSRFDPM-IMJSIDKUSA-N | |
(2S,3S)-1,4-bis(sulfanyl)butane-2,3-diol | |
154.24 | |
CHEBI:42170 | |
≥98.5% | |
Powder |
(1 M soln. in water), (1 M soln. in water) at 0nm, 0.035 max. at 500nm | |
White | |
0.99 | |
HSCH2CH(OH)CH(OH)CH2SH | |
01, III, 2360 | |
dithiothreitol, dl-1,4-dithiothreitol, dl-dithiothreitol, 1,4-dithio-dl-threitol, d-1,4-dithiothreitol, d-dtt, 2s,3s-1,4-dimercaptobutane-2,3-diol, threo-1,4-dimercapto-2,3-butanediol, 1,4-dithiothreitol, dtt | |
C(C(C(CS)O)O)S | |
99% min. (c=5, 660nm, water) 1cm cell | |
446094 | |
154.24 | |
Biochemical | |
DL-1, 4-Dithiothreitol |
Safety and Handling
Causes serious eye irritation, Harmful if swallowed, May cause respiratory irritation, May form combustible dust concentrations in air, Causes skin irritation
Serious eye damage/eye irritation (category 2), Acute toxicity (category 4), Specific target organ toxicity after single exposure (category 3), Skin corrosion/irritation (category 2)
- DL-1,4-Dithiothreitol
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
RUO – Research Use Only